Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Stratifin (14-3-3 sigma), recombinant human protein | 97.0% | 20UG
SDP

Catalog No. 5000177187
Encompass_Preferred

Recombinant human stratifin (14-3-3 sigma) expressed in Escherichia coli and supplied as a lyophilized protein with N-terminal His and C-terminal Myc tags for research use.

  • Recombinant human protein with N-terminal His and C-terminal Myc tags.
  • Expressed in Escherichia coli.
  • Purity 97.0% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 34 kDa.
  • Lyophilized formulation for long-term stability.
  • Low endotoxin level <1 EU/μg.
  • Recommended reconstitution concentration ≥100 μg/mL in ddH2O.
  • Store at -20°C; stable for two years from date of receipt.

Catalog No. 50-001-77187 Supplier Medchemexpress LLC Supplier No. HYP70030820UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human stratifin (14-3-3 sigma) expressed in Escherichia coli and supplied as a lyophilized protein with N-terminal His and C-terminal Myc tags for research use.

  • Recombinant human protein with N-terminal His and C-terminal Myc tags.
  • Expressed in Escherichia coli.
  • Purity 97.0% as determined by reducing SDS-PAGE.
  • Molecular weight approximately 34 kDa.
  • Lyophilized formulation for long-term stability.
  • Low endotoxin level <1 EU/μg.
  • Recommended reconstitution concentration ≥100 μg/mL in ddH2O.
  • Store at -20°C; stable for two years from date of receipt.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.