Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Stratifin protein, human (N-His, C-Myc) | 97.0% | 500UG
SDP

Catalog No. 5000177189
Encompass_Preferred

Recombinant human Stratifin (14-3-3 sigma) is supplied as an N-terminal His and C-terminal Myc tagged lyophilized protein expressed in Escherichia coli for research use. The product is characterized by high purity and defined handling, reconstitution, and storage recommendations.

  • Purity 97.0%.
  • Approximate molecular weight 34 kDa.
  • N-terminal His and C-terminal Myc tags facilitate detection and purification.
  • Lyophilized formulation for long-term stability.
  • Reconstitution recommended at not less than 100 μg/mL.
  • Store lyophilized at -20°C; aliquot and freeze at -20°C or -80°C after reconstitution for extended storage.

Catalog No. 50-001-77189 Supplier Medchemexpress LLC Supplier No. HYP700308500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Stratifin (14-3-3 sigma) is supplied as an N-terminal His and C-terminal Myc tagged lyophilized protein expressed in Escherichia coli for research use. The product is characterized by high purity and defined handling, reconstitution, and storage recommendations.

  • Purity 97.0%.
  • Approximate molecular weight 34 kDa.
  • N-terminal His and C-terminal Myc tags facilitate detection and purification.
  • Lyophilized formulation for long-term stability.
  • Reconstitution recommended at not less than 100 μg/mL.
  • Store lyophilized at -20°C; aliquot and freeze at -20°C or -80°C after reconstitution for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.