Learn More
Medchemexpress LLC Stratifin protein, human (N-His, C-Myc) | 97.0% | 5UG
SDP
Catalog No. 5000177195
Encompass_Preferred
Recombinant human Stratifin (14-3-3 sigma) is provided as a characterized, affinity-tagged protein for use in biochemical assays, antibody validation, and protein-protein interaction studies. The protein is expressed in E. coli and supplied with a Certificate of Analysis documenting purity and molecular weight measurements.
  • Recombinant human protein expressed in E. coli.
  • N-terminal His and C-terminal Myc affinity tags.
  • High purity, ~97.0% by reducing SDS-PAGE.
  • Approximate molecular weight 34 kDa.
  • Stable when stored at -20°C; aliquot and freeze for long-term storage.
Catalog No. 50-001-77195 Supplier Medchemexpress LLC Supplier No. HYP7003085UG
Recombinant human Stratifin (14-3-3 sigma) is provided as a characterized, affinity-tagged protein for use in biochemical assays, antibody validation, and protein-protein interaction studies. The protein is expressed in E. coli and supplied with a Certificate of Analysis documenting purity and molecular weight measurements.
  • Recombinant human protein expressed in E. coli.
  • N-terminal His and C-terminal Myc affinity tags.
  • High purity, ~97.0% by reducing SDS-PAGE.
  • Approximate molecular weight 34 kDa.
  • Stable when stored at -20°C; aliquot and freeze for long-term storage.