Learn More
SPECIALIZEDRX STRAWBERRY WS NAT LIQUID VEGAN
Catalog No. NC3863029
Encompass
A natural / anhydrous / water soluble/miscible liquid flavor concentrate.Contains no allergensGluten-freeSuggested concentration range: 0.1-0.5 percentTaste and aromaVegan compliant /contains no animal products/24-month shelf life from date of manufacture when stored in original, unopened container in a cool dry place.
Catalog No. NC3863029 Supplier SPECIALIZEDRX Supplier No. F014290
A natural / anhydrous / water soluble/miscible liquid flavor concentrate.Contains no allergensGluten-freeSuggested concentration range: 0.1-0.5 percentTaste and aromaVegan compliant /contains no animal products/24-month shelf life from date of manufacture when stored in original, unopened container in a cool dry place.