Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roylco™ Straws & Connectors™
SDPSCI_ED

Catalog No. S13225
Change view
Click to view available options
Includes:
232 Straws, 168 Connectors, Guide
390 Straws, 315 Connectors, Guide
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S13225 390 Straws, 315 Connectors, Guide
S13217 232 Straws, 168 Connectors, Guide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S13225 Supplier Roylco™ Supplier No. R6090
Add to Cart
Add to Cart

Make playhouses and forts, design buildings, and engineer space crafts with these straw and connectors!

  • Durable plastic straws fit snuggly on the connectors, but are flexible so you can build arches and tunnels
  • Appropriate for a wide age range and easy to clean
  • Age: 4+
  • Made in USA

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade 1+
Format Kit
Includes 390 Straws, 315 Connectors, Guide
Quantity 705 Pieces
Languages English, Spanish, French, German
Subject STEM, Physics - Matter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.