Learn More
Aviva Systems Biology Corp STRBP Antibody - middle region (ARP40914_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40914_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STRBP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human STRBP. Alternate names: DKFZp434N214, FLJ11307, FLJ14223, FLJ14984, MGC21529, MGC3405, SPNR, p74, ILF3LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-57661