Learn More
ABclonal Technology STRBP Rabbit pAb
SDP
Catalog No. 5000108531
Encompass_Preferred
Enables RNA binding activity. Predicted to be involved in spermatogenesis. Predicted to act upstream of or within mechanosensory behavior and spermatid development. Located in nucleus.
Catalog No. 50-001-08531 Supplier ABclonal Technology Supplier No. A15169500UL
Enables RNA binding activity. Predicted to be involved in spermatogenesis. Predicted to act upstream of or within mechanosensory behavior and spermatid development. Located in nucleus.