Learn More
Cole-Parmer Argos Technologies Calibrated Inoculating Streaker Loops, 10 ul, 4 mm ID; 5/PK
Catalog No. 501960660 Shop All Cole-Parmer Products
Encompass_Preferred
These loops attached to a light-weight aluminum handle are made of high quality twisted nichrome wire. The handles are insulated to protect the hand, during use. The loops have been designed to slide along agar surfaces without cutting or tearing. Loops and handles cool rapidly, after sterilization. They are offered calibrated (1l & 10l) or non-calibrated. Non-calibrated loops and handles are available as small, medium or large and color coded for easy identification.
Catalog No. 50-196-0660 Supplier Cole-Parmer Supplier No. 0439646
These loops attached to a light-weight aluminum handle are made of high quality twisted nichrome wire. The handles are insulated to protect the hand, during use. The loops have been designed to slide along agar surfaces without cutting or tearing. Loops and handles cool rapidly, after sterilization. They are offered calibrated (1l & 10l) or non-calibrated. Non-calibrated loops and handles are available as small, medium or large and color coded for easy identification.