Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ 3AA Haz-Lo™ Headlamp

Catalog No. 19819890 Shop All Streamlight Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-819-890 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-819-890 Supplier Streamlight™ Supplier No. 61200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 3AA HAZ-LO is an intrinsically safe long-running headlamp for use in atmospheres containing hazardous gases, vapors and liquids.

  • Impact resistant engineering resin case offering exceptional durability and weather resistance
  • All openings are O-ring sealed
  • Unbreakable polycarbonate lens
  • One (1) Watt Super High Flux Luxeon LED, 50,000 hour lifetime
  • Run time: Up to 11 hours of declining output to 25-Percent intensity

Specifications

Light Source C4 LED
Weight (English) 4.5 oz.
Weight (Metric) 0.128 kg
Battery Type 3 AA Alkaline
Certifications/Compliance UL, C-UL Classified Class 1, Division 1, Groups A,B,C,D, Class II, Division 1, Groups E,F,G Class III, T4
Color Yellow
Controller Type Push-button switch
Dimensions (L x W) 7.82 x 6.3 cm (3.08 x 2.5 in.)
Includes Rubber hard hat strap and elastic strap
Length (English) 3.08 in.
Length (Metric) 7.82 cm
Lens Material Polycarbonate
Width (English) 2.5 in.
Width (Metric) 6.3 cm
Product Type Headlamp
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.