Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ 4AA ProPolymer™ Lux Flashlight

Catalog No. 19500558 Shop All Streamlight Products
Change view
Click to view available options
Battery Run Time:
155 hr.
6 hr.
Weight (English):
6.3 oz.
7.16 oz.
Lumen:
100 lm
67 lm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Battery Run Time Weight (English) Lumen
19-500-558 6 hr. 7.16 oz. 100 lm
19-230-248 155 hr. 6.3 oz. 67 lm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-500-558 Supplier Streamlight™ Supplier No. 68602
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Wrist lanyard

The 4AA ProPolymer Lux is a durable polymer flashlight that provides a longer reaching beam and is ergonomically shaped for maximum hand comfort.

High performance 4AA flashlight features a body constructed of a polymer engineering resin that is virtually indestructible, shock resistant, non-conductive and lightweight.

  • One C4™ LED with a 50,000 hour lifetime
  • Deep reflector produces long-range brilliance and smooth beam
  • 4200 candela, 100 lumens
  • Push-button tail switch for one-handed operation
  • Impact-resistant polymer body
  • Unbreakable polycarbonate lens with scratch resistant coating
  • O-ring sealed, water resistant
  • Corrosion proof and non-conductive
  • Powered by four AA alkaline batteries, included
  • Four hours at full intensity, two additional hours of declining usable light
  • Power regulated for maximum output throughout battery life

Certifications

UL listed for Class I, Division 1; Class II, Division 1; and Class III hazardous locations

Specifications

Light Source C4 LED
Weight (English) 7.16 oz.
Weight (Metric) 0.216 kg
Battery Run Time 6 hr.
Candle Power 4200 cd
Color Yellow or Black
For Use With (Application) For maximum hand comfort
Includes Wrist lanyard connection
Lumen 100 lm
Material Polymer
Product Type Flashlight
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.