Learn More
MSC Streamlight Dualie 2AA Flashlight, Intrinsically Safe, 175 Lumens, 1 ea
Catalog No. 5000766713
Encompass
The Dualie 2AA is a compact, intrinsically safe, multi-function flashlight designed to illuminate both the path ahead and the immediate work area, helping to prevent slips, trips, and falls.
  • Three lighting modes: spot, flood, and spot/flood combo.
  • Dual beams intersect for optimal path illumination and no blind spots.
  • Dual head switches for easy beam selection and one-handed operation.
  • Durable, non-conductive, corrosion-proof polymer resin construction.
  • Rocky stipple texture for a secure grip.
  • Unbreakable polycarbonate face cap and side lens with scratch-resistant coating.
  • IPX4 water-resistant and 2 meter impact resistance tested.
  • Integrated magnet on tail end for hands-free use.
  • Includes two AA alkaline batteries.
  • Intrinsically safe for hazardous environments.
Catalog No. 50-007-66713 Supplier MSC Supplier No. 55615942
The Dualie 2AA is a compact, intrinsically safe, multi-function flashlight designed to illuminate both the path ahead and the immediate work area, helping to prevent slips, trips, and falls.
  • Three lighting modes: spot, flood, and spot/flood combo.
  • Dual beams intersect for optimal path illumination and no blind spots.
  • Dual head switches for easy beam selection and one-handed operation.
  • Durable, non-conductive, corrosion-proof polymer resin construction.
  • Rocky stipple texture for a secure grip.
  • Unbreakable polycarbonate face cap and side lens with scratch-resistant coating.
  • IPX4 water-resistant and 2 meter impact resistance tested.
  • Integrated magnet on tail end for hands-free use.
  • Includes two AA alkaline batteries.
  • Intrinsically safe for hazardous environments.