Learn More
MSC Streamlight Handheld Flashlight, LED, 65 Lumens, Intrinsically Safe, 1 ea
Catalog No. 5000765487
Encompass
This small handheld flashlight is durable, waterproof, and intrinsically safe. It offers a long-lasting 24 hr run time and utilizes affordable, easy-to-find alkaline batteries. With a light output of 65 lumens and a beam distance of 96 meters, it is safety rated for Class I, Div. 1 hazardous locations and IP67 rated for dust-tight and waterproof to 1 meter for 30 minutes.
  • Durable, impact-resistant polymer construction
  • Unbreakable polycarbonate lens
  • Waterproof to 1 meter for 30 minutes; IPX7 rated design
  • Intrinsically safe with Class 1, Div 1 safety rating
  • Light output of 65 lumens
  • Beam distance of 96 meters
  • Uses two AA alkaline batteries for 24 hr run time
  • Compact size at approximately 6-⅓ in. length
Catalog No. 50-007-65487 Supplier MSC Supplier No. 37285491
This small handheld flashlight is durable, waterproof, and intrinsically safe. It offers a long-lasting 24 hr run time and utilizes affordable, easy-to-find alkaline batteries. With a light output of 65 lumens and a beam distance of 96 meters, it is safety rated for Class I, Div. 1 hazardous locations and IP67 rated for dust-tight and waterproof to 1 meter for 30 minutes.
  • Durable, impact-resistant polymer construction
  • Unbreakable polycarbonate lens
  • Waterproof to 1 meter for 30 minutes; IPX7 rated design
  • Intrinsically safe with Class 1, Div 1 safety rating
  • Light output of 65 lumens
  • Beam distance of 96 meters
  • Uses two AA alkaline batteries for 24 hr run time
  • Compact size at approximately 6-⅓ in. length