Learn More
MSC Streamlight Headlamp, 120 Lumen LED, 3 Modes, AAA, 1 ea
Catalog No. 5000686189
Encompass
The Septor® Headlamp is designed for tasks requiring close-range illumination, providing a soft flood light. It features seven white 5mm LEDs with three distinct lighting modes to optimize for brightness or extended run times.
  • Features 7 white 5mm LEDs with three lighting modes
  • Provides soft flood light for close-range tasks
  • Low-level battery indicator (red LED flashes when low)
  • Features a 90° tilting head with ratcheting system to reduce neck fatigue
  • Recessed push-button switch, easy to use with gloved hands
  • IPX4 water resistance and 2-meter impact resistance tested
  • Powered by three AAA alkaline batteries (included)
  • Durable, co-molded rubber body construction
  • Includes elastic headstrap and rubber hard hat strap
Catalog No. 50-006-86189 Supplier MSC Supplier No. 64506496
The Septor® Headlamp is designed for tasks requiring close-range illumination, providing a soft flood light. It features seven white 5mm LEDs with three distinct lighting modes to optimize for brightness or extended run times.
  • Features 7 white 5mm LEDs with three lighting modes
  • Provides soft flood light for close-range tasks
  • Low-level battery indicator (red LED flashes when low)
  • Features a 90° tilting head with ratcheting system to reduce neck fatigue
  • Recessed push-button switch, easy to use with gloved hands
  • IPX4 water resistance and 2-meter impact resistance tested
  • Powered by three AAA alkaline batteries (included)
  • Durable, co-molded rubber body construction
  • Includes elastic headstrap and rubber hard hat strap