Learn More
MSC Streamlight Helmet Light, 350 Lumens, 2 Modes, Rechargeable, 1 ea
Catalog No. 5000792983
Encompass
This helmet light is a reliable hands-free lighting tool designed for fire helmets. It is compact, powerful, and durable, easily attaching above or below the brim of fire helmets. It features a blue taillight LED for visibility in smoke, providing 350 lumens and running for 2.25 hours.
  • Designed as a hands-free lighting tool for fire helmets
  • Compact, powerful, and virtually indestructible
  • Low-profile design for easy helmet attachment
  • Blue taillight LED for visibility in smoke
  • Provides 350 lumens with 2.25 hours run time
  • Large push-button switch for gloved-hand operation
  • SAFE OFF feature prevents accidental activation
  • Lightweight, under 4 oz.
  • Durable aluminum construction withstands heat and smoke
  • IPX7 waterproof to 1 meter for 30 minutes
Catalog No. 50-007-92983 Supplier MSC Supplier No. 98069925
This helmet light is a reliable hands-free lighting tool designed for fire helmets. It is compact, powerful, and durable, easily attaching above or below the brim of fire helmets. It features a blue taillight LED for visibility in smoke, providing 350 lumens and running for 2.25 hours.
  • Designed as a hands-free lighting tool for fire helmets
  • Compact, powerful, and virtually indestructible
  • Low-profile design for easy helmet attachment
  • Blue taillight LED for visibility in smoke
  • Provides 350 lumens with 2.25 hours run time
  • Large push-button switch for gloved-hand operation
  • SAFE OFF feature prevents accidental activation
  • Lightweight, under 4 oz.
  • Durable aluminum construction withstands heat and smoke
  • IPX7 waterproof to 1 meter for 30 minutes