Learn More
Description
Includes
120V AC cord, 12V DC cord, and mounting rack
Automatically turns on when power goes off and turns off when power is restored.
- SMART™ power failure circuitry automatically goes on when the power goes off
- Portable, powerful, industrial-duty, rechargeable lantern
- 8 watt high intensity halogen bi-pin bulb
- 8 watt spot, 80,000 peak beam cp, 150 lumens
- Up to nine hours run time per charge
- 6V sealed lead acid battery
Compliance
Rack meets NFPA 1901-10-1.7 standards.
Specifications
Specifications
| Wattage | 8 W |
| Certifications/Compliance | NFPA 1901-10-1.7 |
| Color | Orange |
| Includes | 120 V AC cord, 12 V DC cord, and mounting rack |
| Lumen | 150 lm |
| Product Type | Power Failure System |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.