Learn More
MSC Streamlight Piggyback Charger, USB, Strion Series, 1 ea
Catalog No. 5000766720
Encompass
This piggyback charger is designed to charge both a Strion series flashlight and a spare battery simultaneously. It features a clamp-style holder with a digital control circuit to prevent over-charge and an LED indicator for charge status. The charger can be mounted to a wall or in a vehicle and offers dual power input via a micro USB port or a 2-prong port.
  • Compatible with Strion series flashlights
  • Dual power input with integrated micro USB port and 2-prong port
  • Two-prong port accepts 22051 DC-car cord, 22311 AC-home cord, and 22060 AC-home cord
  • Full charge for flashlight and spare battery takes 6 hours
  • Charging spare battery only takes 3 hours
  • Digital control circuit prevents over-charge
  • LED indicator shows charge status
  • Mountable to a wall or in a vehicle
  • Includes charger base only; battery and cords sold separately
Catalog No. 50-007-66720 Supplier MSC Supplier No. 54840392
This piggyback charger is designed to charge both a Strion series flashlight and a spare battery simultaneously. It features a clamp-style holder with a digital control circuit to prevent over-charge and an LED indicator for charge status. The charger can be mounted to a wall or in a vehicle and offers dual power input via a micro USB port or a 2-prong port.
  • Compatible with Strion series flashlights
  • Dual power input with integrated micro USB port and 2-prong port
  • Two-prong port accepts 22051 DC-car cord, 22311 AC-home cord, and 22060 AC-home cord
  • Full charge for flashlight and spare battery takes 6 hours
  • Charging spare battery only takes 3 hours
  • Digital control circuit prevents over-charge
  • LED indicator shows charge status
  • Mountable to a wall or in a vehicle
  • Includes charger base only; battery and cords sold separately