Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ PolyStinger™ LED HAZ-LO™ Safe Rechargeable Flashlight

Catalog No. 19230245 Shop All Streamlight Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-230-245 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-230-245 Supplier Streamlight™ Supplier No. 76412
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The PolyStinger HAZ-LO is an intrinsically safe Class I, Division 1 full-sized handheld flashlight constructed of durable polymer. Developed for hazardous locations, atmospheres and environments including petro-chem, utility, and other heavy industrial applications.

  • 130 lumens on high
  • Switch to low mode for 12 hour runtime
  • NiCd battery is rechargeable up to 1000 times
  • Super tough corrosion resistant polymer construction
  • Unbreakable polycarbonate lense with scratch-resistant coating
  • IPX4 rated for water resistance
  • 3 meter impact resistance tested

Specifications

Light Source LED
Weight (English) 13.50 oz.
Weight (Metric) 382.72 g
Battery Run Time 4.0 hr. (high), 12 hr. (low)
Battery Type NiCad
Beam Distance 245 m
Color Yellow
For Use With (Application) Petrochemical and Heavy Industrial Applications with Hazardous Atmospheres
Length (English) 10.20 in.
Length (Metric) 25.91 cm
Lens Material Polycarbonate
Lumen 130 lm
Material Nylon polymer
Product Type Flashlight
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.