Learn More
Streamlight™ PolyTac™ LED Tactical Light
Catalog No. 19500570 Shop All Streamlight Products
Change view
Click to view available options
For Use With (Equipment):
Firefighting Helmets
Catalog No. For Use With (Equipment)
19-500-570 Firefighting Helmets
1 options
Catalog No. 19-500-570 Supplier Streamlight™ Supplier No. 88854
Add to Cart
Add to Cart
The PolyTac Helmet Lighting Kit includes a PolyTac flashlight, a helmet mounting bracket, and a rubber helmet strap. It's a quick and easy, no tools required method to attach your PolyTac flashlight to almost any helmet. Also fits hard hats.
  • 600 lumens
  • Switch to low mode for 65 hour runtime
  • 195 meter beam distance
  • Compact size (under 5.5 inches) and comfortable textured grip
  • Helmet mount bracket is durable, lightweight aluminum that easily attaches to the brim of a firefighter helmet with no tools required
  • PolyTac flashlight is IPX7 rated waterproof and made of super tough nylon polymer
  • High/Low/Strobe modes
Specifications
Battery Type Lithium batteries
For Use With (Equipment) Firefighting Helmets
Includes PolyTac LED flashlight, helmet strap, helmet mount, and lithium batteries
Lens Material Borofloat glass