Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ ProPolymers™ Luxeon™ LED Flashlights

Catalog No. 191500062 Shop All Streamlight Products
Change view
Click to view available options
Color:
Yellow or Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-150-0062 Yellow or Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-150-0062 Supplier Streamlight™ Supplier No. 68244
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

3C flashlight includes three C; 4AA flashlight supplied with four AA batteries

Ten times brighter than a high-intensity LED flashlight

  • 1w, Luxeon super high-flux, 3500 candlepower LED bulbs (10,000 hr. life)
  • Impact-resistant, waterproof, nonconductive polymer construction
  • Wrist lanyard connection

3C flashlight

  • Runs up to 5.5 hr. at full strength (2 hr. declining light)
  • Pocket clip

4AA flashlight

  • Runs up to 4 hr. at full strength (2 hr. declining light)

Helmet mounting bracket for 4AA model

  • Metal style
  • Can be adjusted to fit traditional- or modern-style helmets

Certifications

UL Class I, Division 2, Groups A, B, C, D; Class II, Divsion 2, Groups F, G; Class III, T5 (3C model), T3C (4AA model).

Specifications

Light Source LED
Weight (English) 7.69 oz.
Weight (Metric) 0.21 kg
Battery Run Time 6 hr.
Battery Type 4 AA Lux 4 AA
Candle Power 3500 cd
Certifications/Compliance UL- Class I and II, Div. 1, Groups C, D, F and G, Class I, Div. 2, Groups A, B, C, D; Class III, T4, MSHA
Color Yellow or Black
Includes White LED and alkaline batteries
Lumen 100 lm
Material Polymer
Product Type Flashlight
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.