Learn More
MSC Streamlight Rechargeable LED Flashlight, 175 Lumens, Intrinsically Safe, 1 ea
Catalog No. 5000686796
Encompass
This Streamlight handheld flashlight features an LED bulb with 175 lumens output and a 15-hour maximum run time. It includes a rechargeable battery and has 4 light modes. Constructed from durable nylon, it is designed for tactical and industrial use.
  • Leading Class I, Div. 1 right angle, clip-on flashlight for firefighters, featuring a slim profile and D-ring for accurate light orientation.
  • Its spring-loaded clip easily attaches to belts and harnesses.
  • Class I, Div 1 rated - Intrinsically Safe.
  • Features 175 lumens and 405 meter beam distance on High mode.
  • Low mode offers a longer runtime of up to 15 hours.
  • High/Low/Flash/Moonlight modes.
  • Easy operation with a push-button switch, even with gloves on.
  • Durable nylon construction and 2 meter impact resistance tested.
  • IPX4 rated for water resistance.
Catalog No. 50-006-86796 Supplier MSC Supplier No. 06017966
This Streamlight handheld flashlight features an LED bulb with 175 lumens output and a 15-hour maximum run time. It includes a rechargeable battery and has 4 light modes. Constructed from durable nylon, it is designed for tactical and industrial use.
  • Leading Class I, Div. 1 right angle, clip-on flashlight for firefighters, featuring a slim profile and D-ring for accurate light orientation.
  • Its spring-loaded clip easily attaches to belts and harnesses.
  • Class I, Div 1 rated - Intrinsically Safe.
  • Features 175 lumens and 405 meter beam distance on High mode.
  • Low mode offers a longer runtime of up to 15 hours.
  • High/Low/Flash/Moonlight modes.
  • Easy operation with a push-button switch, even with gloves on.
  • Durable nylon construction and 2 meter impact resistance tested.
  • IPX4 rated for water resistance.