Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ Replacement Parts For LiteBox™ FireBox™ Rechargeable Lanterns

Catalog No. 19822939 Shop All Streamlight Products
Change view
Click to view available options
Product Type:
Flashlight Kit
PCB Standard Assembly Kit
Streamlight Battery
For Use With (Equipment):
Litebox, FireBox Lanterns
Litebox, FireBox Rechargeable Lanterns
PAR 36 Lamp Assembly
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
19-822-939 Flashlight Kit PAR 36 Lamp Assembly
19-823-887 PCB Standard Assembly Kit Litebox, FireBox Rechargeable Lanterns
19-819-834 Streamlight Battery Litebox, FireBox Lanterns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-822-939 Supplier Streamlight™ Supplier No. 40107
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Light Source Halogen
Color Orange
For Use With (Application) Fire, Emergency and Rescue
For Use With (Equipment) PAR 36 Lamp Assembly
Product Type Flashlight Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.