Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ Stinger™ Flashlight

Catalog No. 19819840 Shop All Streamlight Products
Change view
Click to view available options
Battery Type:
NiCad
Includes:
AC/DC fast charger, 2 holders
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Battery Type Includes
19-819-840 NiCad AC/DC fast charger, 2 holders
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-819-840 Supplier Streamlight™ Supplier No. 75024

missing translation for 'validMainframeMsg'

Compact, rechargeable, and exceptionally bright

  • Batteries rechargeable up to 1000 times; standard charge time is 10 hours
  • Each charge provides approximately 1 hour, 15 minutes of continuous use
  • Maximum output: 11,000 candela; 90 lumens
  • Black knurled aircraft aluminum body with unbreakable polycarbonate lens can easily be carried on a duty belt
  • Store in charging base for maximum strength charge at all times

Order Info

Length: 7.38 in. (18.7cm). Weight: 10 oz. (284g).

Certifications

UL listed for certain Class I and II, Division 2 hazardous locations; CE marked.

Specifications

Gas Fill Xenon
Weight (English) 10 oz.
Weight (Metric) 0.283 kg
Battery Run Time 1 hr.
Battery Type NiCad
Certifications/Compliance CE, UL Class I, Div. 2, Groups A,B,C,D T1, Class II, Div. 2, Groups F,G T3
Includes AC/DC fast charger, 2 holders
Length (English) 7.38 in.
Length (Metric) 18.7 cm
Lens Material Polycarbonate
Material Aluminum
Product Type Rechargeable Flashlight
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.