Learn More
Description
- Batteries rechargeable up to 1000 times; standard charge time is 10 hours
- Each charge provides approximately 1 hour, 15 minutes of continuous use
- Maximum output: 11,000 candela; 90 lumens
- Black knurled aircraft aluminum body with unbreakable polycarbonate lens can easily be carried on a duty belt
- Store in charging base for maximum strength charge at all times
Order Info
Length: 7.38 in. (18.7cm). Weight: 10 oz. (284g).
Certifications
UL listed for certain Class I and II, Division 2 hazardous locations; CE marked.
Specifications
Specifications
| Gas Fill | Xenon |
| Weight (English) | 10 oz. |
| Weight (Metric) | 0.283 kg |
| Battery Run Time | 1 hr. |
| Battery Type | NiCad |
| Certifications/Compliance | CE, UL Class I, Div. 2, Groups A,B,C,D T1, Class II, Div. 2, Groups F,G T3 |
| Includes | AC/DC fast charger, 2 holders |
| Length (English) | 7.38 in. |
| Length (Metric) | 18.7 cm |
| Lens Material | Polycarbonate |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.