Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck A1c-Cellular™ HbA1c Control
GSA_VASDP

Catalog No. 23045422 Shop All Streck Laboratories Products
Change view
Click to view available options
Quantity:
2 x 2 mL
6 x 2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-045-422 6 x 2 mL
23-045-423 2 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-045-422 Supplier Streck Supplier No. 211124
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

With A1c-Cellular, you can evaluate the entire HbA1c analysis process, from instrument to reagents, to ensure accurate patient results.

  • Available in two clinically significant levels.
  • Appropriate for immunoassay and ionic exchange HPLC methodologies.
  • Assayed on the top HbA1c analyzers.
  • 30-day open-vial stability, 190-day closed-vial stability.
  • Tests the entire process including the lysis step.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance CE
Content And Storage 2°C to 8°C
Format Cap-pierceable Plastic Vial
For Use With (Equipment) Abbott Architect™ c system, Menarini ADAMS™ A1c HA-8160, Beckman Coulter™ UniCel™ DxC 600/800, Bio-rad™ D-10/Variant II™/Variant II™
Type A1c-cellular™ Clinical Control
Intended Use For evaluating the accuracy and precision of HbA1c procedures
Level 1, 2
Quantity 6 x 2 mL
Source Human RBC
Stability 180 Days
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.