Learn More
Description
- Assayed on the Abbott Ruby instrument.
- Plastic vials with pierceable caps for analyzer autosampling.
- 5-day open-vial stability, 45-day closed-vial stability.
- Helps clinical laboratories meet quality requirements.
- Established assay ranges provide less room for error.
Specifications
Specifications
| For Use With (Equipment) | Abbott CELL-DYN™ 1700, 3000, 3200, 3500 and 3700 analyzer |
| Type | Cal-Chex A Plus Hematology Calibrators |
| Quantity | 1 x 3 mL |
| Stability | 5 Days Open, 45 Days Closed Vial |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
