Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck Cal-Chex A Plus Calibrator
SDP

Catalog No. 23285732 Shop All Streck Laboratories Products
Change view
Click to view available options
Quantity:
1 x 3 mL
2 x 3 mL
3 x 3 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-285-732 3 x 3 mL
23-285-729 1 x 3 mL
23-285-730 2 x 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-285-732 Supplier Streck Supplier No. 221108

missing translation for 'validMainframeMsg'

Cal-Chex A Plus is a whole blood product assayed for the calibration of the Abbott™ Ruby™ multi-parameter hematology analyzer.

  • Assayed on the Abbott Ruby instrument.
  • Plastic vials with pierceable caps for analyzer autosampling.
  • 5-day open-vial stability, 45-day closed-vial stability.
  • Helps clinical laboratories meet quality requirements.
  • Established assay ranges provide less room for error.

Specifications

For Use With (Equipment) Abbott CELL-DYN™ 1700, 3000, 3200, 3500 and 3700 analyzer
Type Cal-Chex A Plus Hematology Calibrators
Quantity 3 x 3 mL
Stability 5 Days Open, 45 Days Closed Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.