Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck CD-Chex™ Plus Immunophenotyping Control
SDP

Catalog No. 23046503A Shop All Streck Laboratories Products
Change view
Click to view available options
Level:
CD4 Low
CD4 Low, Normal
Normal
Quantity:
2 x 3 mL
4 x 3 mL
5 x 3 mL
10 x 3 mL
Format:
Cap-pierceable Plastic Vial
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level Quantity Format
23-046-503A Normal 2 x 3 mL Cap-pierceable Plastic Vial
23-046-503B CD4 Low 2 x 3 mL Cap-pierceable Plastic Vial
23-046-503C Normal 5 x 3 mL Cap-pierceable Plastic Vial
23-046-503D CD4 Low 5 x 3 mL Cap-pierceable Plastic Vial
23-046-503F CD4 Low, Normal 10 x 3 mL Cap-pierceable Plastic Vial
23-046-503E CD4 Low, Normal 4 x 3 mL Cap-pierceable Plastic Vial
23-285-420 CD4 Low, Normal 2 x 3 mL Cap-pierceable Plastic Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 23-046-503A Supplier Streck Supplier No. 213365
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CD-Chex Plus is a positive procedural control for T-lymphocyte, B-lymphocyte and NK cell enumeration and features two clinically relevant levels of CD4+ cells, a normal level of CD34+ cells and several other parameters for use as leukemia and lymphoma quality controls.

  • Assayed for 30 parameters, including T-lymphocytes, B-lymphocytes and NK cells.
  • Contains a CD34+ population and is available in two clinically relevant CD4+ levels.
  • Compatible with BD Biosciences and Beckman Coulter flow cytometry systems.
  • 30-day open-vial stability, 90-day closed-vial stability.
  • Manufacturer established assay ranges – no need to verify range per CAP requirements.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.

Specifications

Certifications/Compliance CE
Content And Storage 2°C to 10°C
Format Cap-pierceable Plastic Vial
For Use With (Equipment) Beckman Coulter™ flow cytometry system
Type CD-Chex™ plus Immunophenotyping Control
Intended Use Immunophenotyping including RBC lysis, Antibody staining, instrument performance and operator technique
Level Normal
Quantity 2 x 3 mL
Source Human Peripheral Blood Leukocytes and Erythrocytes
Stability 3 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.