Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck CD-Chex™ Select Immunophenotyping Controls
SDP

Catalog No. 23045501 Shop All Streck Laboratories Products
Change view
Click to view available options
Quantity:
2 x 3 mL
4 x 3 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-045-501 4 x 3 mL
23-045-500 2 x 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-045-501 Supplier Streck Supplier No. 213555

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CD-Chex Select is a positive flow cytometry control for CD26, CD41, CD57, CD58, CD61, CD64, CD79a, FMC7, MPO, TCR α/β and TCR γ/δ expression.

  • Assayed for 11 CD markers, including 2 intracellular markers.
  • Whole blood control resembles patient sample.
  • Compatible with automated sample preparation instruments and BD Biosciences and Beckman Coulter flow cytometry systems.
  • 30-day open-vial stability, 90-day closed-vial stability.
  • Manufacturer established assay ranges – no need to verify range per CAP requirements.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Content And Storage 2°C to 8°C
Format Vial
For Use With (Equipment) BD biosciences and Beckman Coulter™ flow cytometry system
Type CD-Chex™ Select Flow Cytometry Control
Intended Use Reagent staining, Erythrocyte lysis, sample preparation, instrument performance, immunophenotyping
Quantity 4 x 3 mL
Source Stabilized Human Blood
Stability 3 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.