Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck Cell-Chex™ Cell Body Fluid Control
GSA_VASDP

Catalog No. 11716331 Shop All Streck Laboratories Products
Change view
Click to view available options
Level:
1- UC
1- UC, 2
1- UC/CC, 2
1-CC
2
Quantity:
2 x 2 mL
4 x 2 mL
6 x 2 mL
12 x 2 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level Quantity
11-716-331 1- UC, 2 4 x 2 mL
11-716-345 2 6 x 2 mL
23-285-777 1- UC, 2 12 x 2 mL
23-285-781 1-CC 2 x 2 mL
23-285-782 1-CC 6 x 2 mL
23-285-783 1- UC/CC, 2 2 x 2 mL
23-285-784 1- UC 6 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 11-716-331 Supplier Streck Supplier No. 212420
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cell-Chex is a multi-level manual cerebrospinal and body fluid control for evaluating the accuracy and precision of hemacytometer RBC and WBC counts, urate and CPPD crystal identification and manual WBC differential procedures.

  • Assayed for the 5-part WBC differential.
  • Contains human white blood cells and CPPD crystals.
  • 30-day open-vial stability, 6-month closed-vial stability.
  • Validates the entire cell qualification and quantification procedure.
  • Long shelf life reduces the number of shipments and associated costs.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 10°C
For Use With (Equipment) A hemacytometer or WBC differential counts using a cytocentrifuge smear
Type Cell Body Fluid Control
Intended Use Body fluid procedural control for RBC and WBC counts, crystal identification and WBC differentiation
Level 1- UC, 2
Quantity 4 x 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.