Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck Cell Preservative™
GSA_VASDP

Catalog No. 23046501G Shop All Streck Laboratories Products
Change view
Click to view available options
Quantity:
6 x 1 mL
2 x 10 mL
50 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-501G 50 x 1 mL
23-046-501D 6 x 1 mL
23-045-412 2 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-046-501G Supplier Streck Supplier No. 213355
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell Preservative is an easy-to-use liquid preservative that maintains white blood cell integrity and allows CD markers to be measured through flow cytometric analysis. Cell Preservative stabilizes peripheral and cord blood samples, surgical tissue samples, bone marrow and fine needle aspirates for up to 7 days prior to analysis by flow cytometry.

  • Stabilizes samples for up to 7 days prior to analysis by flow cytometry.
  • Preserves peripheral and cord blood samples, surgical tissue samples, bone marrow and fine needle aspirates.
  • Available in 1 mL and 10 mL vials.
  • Limits the adverse effects of time, storage and transport conditions on sample integrity.
  • Reduces the need for immediate sample processing and analysis.
  • Allows for the batching of samples, optimizing laboratory workflow.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Color Clear
Content And Storage 18°C to 30°C
For Use With (Equipment) Flow cytometer
Type Cell Preservative
Intended Use Easy to use liquid preservative that stabilizes cell morphology and preserves cellular Antigen expression of WBC for up to 7 days
pH ≈7.2
Quantity 50 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.