Learn More
Description
- Provides assay values for the most popular three- and five-part hematology instruments.
- Customized kits with WBC, RBC, Hgb and platelet ranges for each instrument model.
- Plastic vials with pierceable caps for analyzer autosampling.
- 5-day open-vial stability, 120-day closed-vial stability.
- Tests the extreme upper and lower limits of an instrument.
- Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.
Specifications
Specifications
| For Use With (Equipment) | CELL DYN 3000 |
| Type | Linearity Control Kit |
| Includes | WBC, platelet and RBC or Hemoglobin vials |
| Intended Use | CVA for WBC, Platelets, HgB |
| Level | 5 |
| Quantity | 12 x 3 mL |
| Stability | 5 Days Open, 120 Days Closed Vial |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
