Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck CD-Chex™ CD117
SDP

Catalog No. 23045526 Shop All Streck Laboratories Products
Change view
Click to view available options
Packaging:
1 x 0.03 oz. (1 x 1.0 mL)
2 x 0.03 oz. (2 x 1.0 mL)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
23-045-526 1 x 0.03 oz. (1 x 1.0 mL)
23-045-525 2 x 0.03 oz. (2 x 1.0 mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-045-526 Supplier Streck Supplier No. 213558
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CD-Chex CD117 Plus is a flow cytometry control that provides assay values for CD117, CD25 and CD71 on abnormal and total leukocytes.

  • Only commercially available control assayed for CD117, CD25 and CD71.
  • Resembles an abnormal peripheral blood patient sample.
  • 30-day open-vial stability, 90-day closed-vial stability.
    Aids in meeting the quality requirements for leukemia and lymphoma immunophenotyping.
  • Manufacturer established assay ranges – no need to verify range per CAP requirements.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.

Specifications

Content And Storage 2°C to 10°C
For Use With (Equipment) BD Biosciences and Beckman Coulter™ flow cytometry system
Type CD-Chex™ CD117 Plus Cytometry Control
For Use With (Application) Positive procedural control for CD117, CD25 and CD71, to monitor reagent staining, erythrocyte lysis, sample preparation, and instrument performance, eliminates the need for labs to hold and validate patient samples
Packaging 1 x 0.03 oz. (1 x 1.0 mL)
Stability 30 Days Open, 90 Days Closed
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.