Learn More
Streck Cell-Chex™ Auto Body Fluid Cell Count Control
SDP
Catalog No. 23046629 Shop All Streck Laboratories Products
Change view
Click to view available options
Level:
Tri-Level Control Solution
Catalog No. Level
23-046-629 Tri-Level Control Solution
1 options
Catalog No. 23-046-629 Supplier Streck Supplier No. 213479
Cell-Chex Auto is an automated body fluid cell count control for Abbott™, Beckman Coulter™, Siemens Healthineers™ and Sysmex™ hematology analyzers. With this tri-level control, you can verify instrument accuracy at the lower WBC and RBC reportable range limits that standard CBC controls and calibrator material fail to assess.
  • Three levels of control.
  • 30-day open-vial stability, 75-day closed-vial stability.
  • Verifies instrument accuracy at the lower WBC and RBC reportable range limits than standard controls.
  • Can be used to establish an analyzer’s sensitivity.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.
Specifications
Type Cell-Chex™ Auto Body Fluid Cell Count Control
Intended Use Abbott, Beckman Coulter™, Siemens Healthcare Diagnostics and Sysmex™ hematology instruments
Level Tri-Level Control Solution