Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck CD-Chex TdT™ Plus Assay
SDP

Catalog No. 23285546 Shop All Streck Laboratories Products
Change view
Click to view available options
Quantity:
1 x 1 mL
2 x 1 mL
Stability:
30 Days Open
90 Days Open
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Stability
23-285-546 2 x 1 mL 30 Days Open
23-285-545 1 x 1 mL 90 Days Open
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-285-546 Supplier Streck Supplier No. 213566
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CD-Chex TdT Plus is a positive procedural control that provides reference values for TdT, CD1a, CD2, CD4, CD5, CD7, CD8, CD34 and cytoplasmic CD3. This control contains several markers designated as rare flow antigens by CAP.

  • Only flow cytometry control assayed for TdT, CD1a, CD34 and cytoplasmic CD3.
  • Offers an abnormally high level of CD34: over 60% positive for CD34+.
  • Compatible with BD Biosciences and Beckman Coulter flow cytometry systems.
  • 30-day open-vial stability, 90-day closed-vial stability.
  • Manufacturer established assay ranges – no need to verify range per CAP requirements.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.

Specifications

For Use With (Equipment) BD Biosciences and Beckman Coulter flow cytometry systems
Type CD-Chex TdT™ Plus Assay
Quantity 2 x 1 mL
Stability 30 Days Open
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.