Learn More
Streck Para 4™ Assayed Hematology Control
SDP
Catalog No. 23285645 Shop All Streck Laboratories Products
Change view
Click to view available options
Type:
Para 4 Hematology Control
Catalog No. Type
23-285-645 Para 4 Hematology Control
1 options
This item is not returnable. View return policy
Catalog No. 23-285-645 Supplier Streck Supplier No. 215103
This item is not returnable. View return policy
Para 4 is a hematology control for laboratories that perform a limited number of hematology tests utilizing manual or semi-manual methods. It is available in low, normal and high levels.
  • Clear results in 20 minutes – test up to 12 samples per hour.
  • Compatible with standard 13 x 75 mm K2 or K3EDTA tubes and 500 µL BD Microtainer and BD Microtainer MAP EDTA tubes.
  • User-friendly touchscreen interface.
  • External barcode scanner and printer.
  • QC and patient archives store 5,000 results per file.
  • Produces ESR results without consuming patient sample.
Specifications
Type Para 4 Hematology Control