Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck Para™ 5X Whole Blood Control
SDP

Catalog No. 23285572 Shop All Streck Laboratories Products
Change view
Click to view available options
Quantity:
3 x 1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-285-572 3 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-285-572 Supplier Streck Supplier No. 215403

missing translation for 'validMainframeMsg'

Para 4 is a hematology control for laboratories that perform a limited number of hematology tests utilizing manual or semi-manual methods. It is available in low, normal and high levels.

  • Whole blood control resembles patient sample.
  • Available in three clinically significant levels.
  • Packaged in plastic squeeze dropper vials and glass vials.
  • 14-day open-vial stability, 110-day closed-vial stability.
  • Long shelf life reduces the number of shipments and associated costs.
  • Access to STATS™, Streck’s free interlaboratory quality control program with peer group data comparison and online submission.

Order Info

Packaged in a 3.0mL pierceable cap vial compatible with the auto-sampling capabilities of the Pentra 60C+ and 80 and the Ac·T 5diff.

Specifications

Format Glass Vial
For Use With (Equipment) Horiba medical ABX Pentra™ 60C+ and 80 and the Coulter™ Ac
Type Para™ 5X Whole Blood Control
Includes Assay disc
Intended Use White cell differential, positioning the populations within the limitations of the algorithms on the system
Level 1 Low, 1 Normal, 1 High
Quantity 3 x 1.5 mL
Stability 110 Days
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.