Learn More
Streck Pipet Labs Calibration Kit
SDP
Catalog No. 23042701 Shop All Streck Laboratories Products
Change view
Click to view available options
Volume (Metric):
10 to 200 μL
201 to 5000 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
23-042-701 201 to 5000 μL
23-042-700 10 to 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-042-701 Supplier Streck Supplier No. 271279
Includes
Six vials of receiver solution and one or two vials of pipetting solution
Pipet Verification Service Kit is a convenient and simple way to verify the calibration of laboratory pipets. PVS kits are ideal for pipets calibrated for volumes from 10 μL to 5000 μL.
  • Easy-to-use kits are shipped to the lab.
  • NIST-traceable.
  • Compatible with any pipette brand.
  • Rapid turnaround.
  • Pipettes do not have to leave the lab.
  • Users receive an email version of a regulatory-compliant report.
Order Info
Order one PVS kit per pipet to be tested
Specifications
Product Type Maintenance Kit
For Use With (Application) To perform simple titrations, analysis and calculation of the results
Volume (Metric) 201 to 5000 μL
Includes Six vials of receiver solution and one or two vials of pipetting solution.
Quantity 1 Ea.