Learn More
Streck Retic-Chex™ II Controls
SDP
Catalog No. p-4932739 Shop All Streck Laboratories Products
Change view
Click to view available options
Level:
1, 2
1, 3
Type:
Hematology II Controls
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level Type
23-285-644 1, 3 Hematology II Controls
23-285-212 1, 2 Hematology II Controls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-285-644 Supplier Streck Supplier No. 285211
Retic-Chex II is a tri-level whole blood control with reticulocytes used to verify the accuracy and precision of manual reticulocyte counting methods.
  • Can be used for manual and automated reticulocyte counting.
  • Available in three clinically significant levels.
  • Packaged in 1.0 mL plastic squeeze dropper vials for accurate dispensing.
  • 14-day open-vial stability, 75-day closed-vial stability.
  • Established assay ranges provide less room for error.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.
Specifications
Type Hematology II Controls
Level 1, 3