Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck Retic-Chex™ II Stains and Controls
SDP

Catalog No. 23046569 Shop All Streck Laboratories Products
Change view
Click to view available options
Level:
1
1, 2, 3
2
3
Quantity:
4 x 1 mL
6 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level Quantity
23-046569 1 4 x 1 mL
23-046570 2 4 x 1 mL
23-046571 3 4 x 1 mL
23-285-106 1, 2, 3 6 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-046569 Supplier Streck Supplier No. 285104

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Retic-Chex II is a tri-level whole blood control with reticulocytes used to verify the accuracy and precision of manual reticulocyte counting methods.

  • Can be used for manual and automated reticulocyte counting.
  • Available in three clinically significant levels.
  • Packaged in 1.0 mL plastic squeeze dropper vials for accurate dispensing.
  • 14-day open-vial stability, 75-day closed-vial stability.
  • Established assay ranges provide less room for error.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.

Specifications

Certifications/Compliance CE
Content And Storage 2°C to 10°C
Format Plastic Squeeze Dropper Vial
For Use With (Equipment) Beckman Coulter™ STKS™, MAXM™, HmX, LH 500, Abbott CELL-DYN™ 3500 and 3700
Type Hematology Control
Intended Use For laboratory quality control procedure requirements
Level 1
Quantity 4 x 1 mL
Source Human Reticulocytes
Stability 1 Year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.