Learn More
Streck Retic-Chex™ Linearity
SDP
Catalog No. 23045404 Shop All Streck Laboratories Products
Change view
Click to view available options
Quantity:
5 x 1 mL
5 x 3 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-045-404 5 x 3 mL
23-042-626 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-045-404 Supplier Streck Supplier No. 285535
Add to Cart
Add to Cart
This item is not returnable. View return policy
Retic-Chex Linearity for BC is designed to establish the reportable range and linearity of Beckman Coulter™ hematology analyzers that report the reticulocyte parameter.
  • Plastic vials with pierceable caps for analyzer autosampling.
  • Assayed for Beckman Coulter hematology analyzers.
  • 5-day open-vial stability, 105-day closed-vial stability.
  • Established assay ranges provide less room for error.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.
Compliance
Complies with CLIA 88 Regulations [42 CFR 493.1217 (b)(2)(ii)(C)], and CAP Sections 02.2576 and 02.2579 for calibration verification.
Specifications
Certifications/Compliance CLIA-88
Type Linearity
Intended Use Calibration verification assessment, establishing the analytic linearity of automated Reticulocyte counting instruments
Level 1 to 5
Quantity 5 x 3 mL
Stability 8 Days Open Vial