Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck STaK-Chex™ Plus Retics Control
SDP

Catalog No. p-2736383 Shop All Streck Laboratories Products
Change view
Click to view available options
Level:
1 Low, 1 Normal, 1 High
12 Normal
4 High
4 Low
4 Low, 4 Normal, 4 High
Quantity:
3 x 4.5 mL
4 x 4.5 mL
12 x 4.5 mL
Format:
Piercable-cap Vial
Vial
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level Quantity Format
23-046-407 12 Normal 12 x 4.5 mL Piercable-cap Vial
23-046-410 4 Low 4 x 4.5 mL Vial
23-046-411 4 High 4 x 4.5 mL Vial
11-716-335 4 Low, 4 Normal, 4 High 12 x 4.5 mL Vial
23-046-408 1 Low, 1 Normal, 1 High 3 x 4.5 mL Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 23-046-407 Supplier Streck Supplier No. 225483
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

STaK-Chex Plus Retics is a comprehensive, whole blood five-part white cell differential control with reticulocytes and nucleated red blood cells for use on Beckman Coulter™ instruments. It features vials with pierceable caps for autosampling and is available in low, normal and high levels.

  • Includes all CBC parameters, reticulocytes and nucleated RBCs.
  • Available in three clinically significant levels.
  • Plastic vials with pierceable caps for analyzer autosampling.
  • Assay values are available for download.
  • 14-day open-vial stability, 105-day closed-vial stability.
  • Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.

Specifications

Certifications/Compliance CE
Content And Storage 2°C to 10°C
Format Piercable-cap Vial
For Use With (Equipment) Beckman Coulter™ UniCel™ DxH™ 800, LH780/785, LH 750/755, LH500 and HmX
Type STaK-Chex™ plus Retics Control
Includes Assay Disc
Intended Use For evaluating the accuracy and precision of automated procedures that measure Blood cell parameters
Level 12 Normal
Quantity 12 x 4.5 mL
Stability 105 Days
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.