Learn More
Grainger Street Tee 316 SS 3/4 in 3500 PSI
Catalog No. 502753125
Encompass_Preferred
The Parker Street Tee 316 Stainless Steel is a 316 stainless steel street tee fitting with female NPT threaded connections. It has a maximum pressure rating of 6000 psi at 100°F and is made in the USA. This street tee fitting allows flow through the run with a branch connection and is commonly used in hydraulic, instrumentation, and industrial applications. The body material provides good corrosion resistance. Key dimensions include 1/4" nominal pipe size, 1/4" NPT female thread connections, and 1-1/4" center to end dimension.
Catalog No. 50-275-3125 Supplier Grainger Supplier No. 14M060
The Parker Street Tee 316 Stainless Steel is a 316 stainless steel street tee fitting with female NPT threaded connections. It has a maximum pressure rating of 6000 psi at 100°F and is made in the USA. This street tee fitting allows flow through the run with a branch connection and is commonly used in hydraulic, instrumentation, and industrial applications. The body material provides good corrosion resistance. Key dimensions include 1/4" nominal pipe size, 1/4" NPT female thread connections, and 1-1/4" center to end dimension.