Learn More
Ey Laboratories Inc Strep-A-Chek Kit, Rapid Detection of Group A Streptococcus, Clear Visual Results, 100 Tests
Catalog No. NC1187109
Encompass
The Strep-A-Chek Kit is a diagnostic assay providing rapid detection of Group A Streptococcus and Enterococci through the identification of pyrrolidonyl arylamidase (PYR) activity. Its simple visual results, distinct pink/red color for positives, and inclusiveness of reagent strips and color developer enhance user convenience. Ideal for trained lab personnel, the kit is designed for timely, efficient presumptive identification, setting it apart from other products.
Catalog No. NC1187109 Supplier Ey Laboratories Inc Supplier No. 1305000
The Strep-A-Chek Kit is a diagnostic assay providing rapid detection of Group A Streptococcus and Enterococci through the identification of pyrrolidonyl arylamidase (PYR) activity. Its simple visual results, distinct pink/red color for positives, and inclusiveness of reagent strips and color developer enhance user convenience. Ideal for trained lab personnel, the kit is designed for timely, efficient presumptive identification, setting it apart from other products.