Learn More
InBios International, Inc. Strep A RespiraDX™ Rapid Test

Description
Strep A RespiraDX™ Rapid Test is for the qualitative detection of Strep A antigen from throat swab specimens to aid in the diagnosis of Group A Streptococcal infection.
- US FDA 510(k) cleared for in vitro diagnostic use
- CLIA-Waived for Point-of-Care use
- Sensitivity: 96.5%, Specificity: 99.5%
- External positive and negative controls included
- Room-temperature storage
Specifications
Specifications
| CE Marker | No |
| Clia Complexity | Waived |
| Detectable Analytes | Streptococci (Group A) |
| DoA Calibrators | No |
| Type | Rapid Test |
| Disposable | Single-use |
| Form | Swab |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.