Learn More
Apexbio Technology LLC Streptavidin Beads T3 5x1ml
SDP
Catalog No. 503107441
Encompass_Preferred
Our Streptavidin Beads T3 magnetic beads are an ideal choice for numerous applications including purification of proteins and nucleic acids protein interaction studies immunoprecipitation assays immunoassays phage display bio-screening drug screening and cell isolation Add the magnetic beads to samples containing biotinylated molecules such as peptides proteins antibodies carbohydrates lectins oligonucleotides DNA or RNA After a brief incubation the biotinylated molecules will bind to the beads and the resulting bead-molecule complexes can then be separated using a magnet The capture washing and detection steps can all be optimized for manual or automated procedures If indirect capture of a target is required first mix the biotinylated molecules with the target samples to form molecule-target complexes and then add magnetic beads to the sample
Catalog No. 50-310-7441 Supplier Apexbio Technology LLC Supplier No. K13015
Our Streptavidin Beads T3 magnetic beads are an ideal choice for numerous applications including purification of proteins and nucleic acids protein interaction studies immunoprecipitation assays immunoassays phage display bio-screening drug screening and cell isolation Add the magnetic beads to samples containing biotinylated molecules such as peptides proteins antibodies carbohydrates lectins oligonucleotides DNA or RNA After a brief incubation the biotinylated molecules will bind to the beads and the resulting bead-molecule complexes can then be separated using a magnet The capture washing and detection steps can all be optimized for manual or automated procedures If indirect capture of a target is required first mix the biotinylated molecules with the target samples to form molecule-target complexes and then add magnetic beads to the sample