Learn More
Carbosynth LLC. Streptavidin Poly-HRP20 Conjugate for use in immunoassays
SDP
Catalog No. 501253505
Encompass_Preferred
Streptavidin Poly-HRP Conjugate is streptavidin biotin-binding protein that is conjugated with polymers of horseradish peroxidase, enabling signal amplification and detection of biotinylated antibodies for ELISA and other methods.
Catalog No. 50-125-3505 Supplier Carbosynth LLC. Supplier No. 85R200100UL
Streptavidin Poly-HRP Conjugate is streptavidin biotin-binding protein that is conjugated with polymers of horseradish peroxidase, enabling signal amplification and detection of biotinylated antibodies for ELISA and other methods.