Learn More
Carbosynth LLC. Streptavidin Poly-HRP40 Conjugate for use in immunoassays
SDP
Catalog No. 501253381
Encompass_Preferred
Streptavidin Poly-HRP Conjugate is streptavidin biotin-binding protein that is conjugated with polymers of horseradish peroxidase, enabling signal amplification and detection of biotinylated antibodies for IHC and other methods. This poly-HRP conjugate is designed to deliver the highest sensitivity and low background in immunoassays where sample volume is limited or when the target molecule is present at low levels. The estimated average number of HRP monomer molecules in SA-PolyHRP20 conjugate is 100 (20 X 5), in SA-PolyHRP40 - 200 (40 X 5) and in SA-PolyHRP80 - 400 (80 X 5). Thus, PolyHRP brings in reaction with substrate development system much larger number of enzyme label molecules (per one bound analyte molecule) than conventional conjugates do.
Catalog No. 50-125-3381 Supplier Carbosynth LLC. Supplier No. 65RS104PHRP200UG
Streptavidin Poly-HRP Conjugate is streptavidin biotin-binding protein that is conjugated with polymers of horseradish peroxidase, enabling signal amplification and detection of biotinylated antibodies for IHC and other methods. This poly-HRP conjugate is designed to deliver the highest sensitivity and low background in immunoassays where sample volume is limited or when the target molecule is present at low levels. The estimated average number of HRP monomer molecules in SA-PolyHRP20 conjugate is 100 (20 X 5), in SA-PolyHRP40 - 200 (40 X 5) and in SA-PolyHRP80 - 400 (80 X 5). Thus, PolyHRP brings in reaction with substrate development system much larger number of enzyme label molecules (per one bound analyte molecule) than conventional conjugates do.