Learn More
Thermo Scientific™ Strep Selective Agar with RNA, Maltose, Neomycin and Polymyxin B
GSA_VA
Catalog No. R01864
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01864 100/Pk.
R01857 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R01864 Supplier Thermo Scientific™ Supplier No. R01864
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Isolate and cultivate Streptococcus pyogenes from throat cultures and upper respiratory specimens with this selective, ready-to-use medium.
Isolate and cultivate Streptococcus pyogenes from throat cultures and upper respiratory specimens with Thermo Scientific™ Remel™ Strep Selective Agar prepared plates with RNA, maltose, neomycin, polymyxin B. The medium is based on the formulation of Roantree, Rantz and Haines1. The addition of ribonucleic acid and maltose increases the production of the streptolysin S, the active substance causing hemolysis on blood agar. Neomycin and polymyxin B are selective agents which facilitate isolation of S. pyogenes by suppressing commensal microbial flora found in the oropharynx.
Streptococcus pyogenes is an important human pathogen that causes a wide variety of acute infections, such as soft tissue infections and pharyngitis; severe life-threatening infections, such as streptococcal toxic shock syndrome; and devastating post-infectious sequelae, such as rheumatic fever and glomerulonephritis.
Use Strep Selective Agar with RNA, maltose, neomycin, polymyxin B for the primary isolation of Streptococcus pyogenes from throat cultures and upper respiratory specimens.
  • Selective: selective agents neomycin and polymyxin B suppress commensal microbial flora, including coliforms, staphylococci, Micrococcus, Haemophilus, and Neisseria
  • Ready-to-use: the convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Strep Selective Agar w/RNA, maltose, neomycin, polymyxin B
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 100/Pk.