Learn More
Zymo Research Corporation Strep-Spin Protein Miniprep Kit (10 preps)
SDP
Catalog No. 502037213
Encompass
Up to 450 g of Strep-tagged protein can be purified in 7 minutes and eluted in Strep-Elution Buffer. The purified protein can be used directly for enzymatic assays, protein biochemical analyses, SDS-PAGE and other applications. The product has been optimized for maximal protein purity. The straightforward spin wash elute protocol dramatically simplifies protein purification: get results in minutes, not hours.
Catalog No. 50-203-7213 Supplier Zymo Research Corporation Supplier No. P2004
Up to 450 g of Strep-tagged protein can be purified in 7 minutes and eluted in Strep-Elution Buffer. The purified protein can be used directly for enzymatic assays, protein biochemical analyses, SDS-PAGE and other applications. The product has been optimized for maximal protein purity. The straightforward spin wash elute protocol dramatically simplifies protein purification: get results in minutes, not hours.