Learn More
IBA LifeSciences Strep-Tactin; 5 mg
Catalog No. 502100136
Encompass
Strep-Tactin is a streptavidin variant, which can be utilized for coating of different surfaces. It is optimized for the binding of Strep-tagII and Twin-Strep-tag fusion proteins but still maintains the ability to capture biotinylated targets. Therefore, it can be used for assay development and protein interaction studies, including experiments such as proximity-dependent biotin identification (BioID).
Catalog No. 50-210-0136 Supplier IBA LifeSciences Supplier No. 21204005
Strep-Tactin is a streptavidin variant, which can be utilized for coating of different surfaces. It is optimized for the binding of Strep-tagII and Twin-Strep-tag fusion proteins but still maintains the ability to capture biotinylated targets. Therefore, it can be used for assay development and protein interaction studies, including experiments such as proximity-dependent biotin identification (BioID).